Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601213 MIM: 613745 | ||||||||||||||||||||
Literature Links: |
ABCE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCE1 - ATP binding cassette subfamily E member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040876.1 | 293 | UTR 5 | NP_001035809.1 | |||
NM_002940.2 | 293 | UTR 5 | NP_002931.2 |
ANAPC10 - anaphase promoting complex subunit 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256706.1 | 293 | Intron | NP_001243635.1 | |||
NM_001256707.1 | 293 | Intron | NP_001243636.1 | |||
NM_001256708.1 | 293 | UTR 5 | NP_001243637.1 | |||
NM_001256709.1 | 293 | Intron | NP_001243638.1 | |||
NM_001256710.1 | 293 | Intron | NP_001243639.1 | |||
NM_001256711.1 | 293 | Intron | NP_001243640.1 | |||
NM_001256712.1 | 293 | Intron | NP_001243641.1 | |||
NM_001318367.1 | 293 | Intron | NP_001305296.1 | |||
NM_014885.4 | 293 | Intron | NP_055700.2 |