Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGTCGTGTGCAATTCCTGGGGC[A/G]CAGGTAGTAAAGGTGTACAGTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601522 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRB7 - growth factor receptor bound protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030002.2 | 745 | Intron | NP_001025173.1 | |||
NM_001242442.1 | 745 | Intron | NP_001229371.1 | |||
NM_001242443.1 | 745 | Intron | NP_001229372.1 | |||
NM_005310.3 | 745 | Intron | NP_005301.2 | |||
XM_017024536.1 | 745 | Silent Mutation | GCA,GCG | A,A 16 | XP_016880025.1 | |
XM_017024537.1 | 745 | Intron | XP_016880026.1 | |||
XM_017024538.1 | 745 | Silent Mutation | GCA,GCG | A,A 16 | XP_016880027.1 |