Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACCTTCTGGAATAGATGATCC[C/G]AAAGGATGACCTTAGGATTCCCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOT12 - acyl-CoA thioesterase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130767.2 | Intron | NP_570123.1 | ||||
XM_006714531.2 | Intron | XP_006714594.1 | ||||
XM_006714532.3 | Intron | XP_006714595.1 | ||||
XM_017009045.1 | Intron | XP_016864534.1 | ||||
XM_017009046.1 | Intron | XP_016864535.1 | ||||
XM_017009047.1 | Intron | XP_016864536.1 | ||||
XM_017009048.1 | Intron | XP_016864537.1 |