Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATGCACTGGGCCTTCTTCACA[A/G]TGAGTAGGTACTGAAGTGACAGTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612538 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC49 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LRRC49 - leucine rich repeat containing 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199017.2 | 671 | Intron | NP_001185946.1 | |||
NM_001199018.2 | 671 | Intron | NP_001185947.1 | |||
NM_001284357.1 | 671 | Intron | NP_001271286.1 | |||
NM_017691.4 | 671 | Intron | NP_060161.2 | |||
XM_005254492.4 | 671 | Intron | XP_005254549.1 | |||
XM_011521715.2 | 671 | Intron | XP_011520017.1 | |||
XM_011521717.2 | 671 | Intron | XP_011520019.1 | |||
XM_011521718.2 | 671 | Intron | XP_011520020.1 | |||
XM_017022357.1 | 671 | Intron | XP_016877846.1 | |||
XM_017022358.1 | 671 | Intron | XP_016877847.1 | |||
XM_017022359.1 | 671 | Intron | XP_016877848.1 |
THAP10 - THAP domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020147.3 | 671 | Silent Mutation | CAC,CAT | H,H 170 | NP_064532.1 |
Set Membership: |
HapMap |