Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGGCACGTCAGTGGCCCAGTCT[C/G]GTGCCCAAGCCCCAGGCAGGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
REPIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
REPIN1 - replication initiator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099695.1 | 218 | Missense Mutation | CGT,GGT | R,G 106 | NP_001093165.1 | |
NM_001099696.2 | 218 | Missense Mutation | CGT,GGT | R,G 49 | NP_001093166.1 | |
NM_013400.3 | 218 | Missense Mutation | CGT,GGT | R,G 49 | NP_037532.2 | |
NM_014374.3 | 218 | Missense Mutation | CGT,GGT | R,G 49 | NP_055189.2 | |
XM_005249985.1 | 218 | Missense Mutation | CGT,GGT | R,G 49 | XP_005250042.1 | |
XM_006715947.3 | 218 | Missense Mutation | CGT,GGT | R,G 106 | XP_006716010.1 | |
XM_006715948.3 | 218 | Missense Mutation | CGT,GGT | R,G 106 | XP_006716011.1 | |
XM_006715949.3 | 218 | Missense Mutation | CGT,GGT | R,G 106 | XP_006716012.1 | |
XM_006715950.3 | 218 | Missense Mutation | CGT,GGT | R,G 105 | XP_006716013.1 | |
XM_006715951.2 | 218 | Missense Mutation | CGT,GGT | R,G 105 | XP_006716014.1 | |
XM_006715952.2 | 218 | Missense Mutation | CGT,GGT | R,G 49 | XP_006716015.1 | |
XM_006715953.2 | 218 | Missense Mutation | CGT,GGT | R,G 49 | XP_006716016.1 | |
XM_011516112.1 | 218 | Missense Mutation | CGT,GGT | R,G 49 | XP_011514414.1 | |
XM_017012081.1 | 218 | Missense Mutation | CGT,GGT | R,G 49 | XP_016867570.1 | |
XM_017012082.1 | 218 | Missense Mutation | CGT,GGT | R,G 49 | XP_016867571.1 |
ZNF775 - zinc finger protein 775 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |