Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGGGAGGCATGAATCCGGGCCG[C/T]GGGGGAGTCTGTTAGGCAAAAGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609744 | ||||||||||||||||||||
Literature Links: |
CADM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CADM4 - cell adhesion molecule 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145296.1 | 631 | Missense Mutation | ACG,GCG | T,A 225 | NP_660339.1 | |
XM_005258620.2 | 631 | Missense Mutation | ACG,GCG | T,A 225 | XP_005258677.1 | |
XM_017026452.1 | 631 | Missense Mutation | ACG,GCG | T,A 206 | XP_016881941.1 |
ZNF428 - zinc finger protein 428 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |