Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCCCCGCCTGGTGGCAGCCCCT[C/T]TCTGCGTCCCTCGCGGCCTGGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616533 MIM: 604892 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX31 - DEAD-box helicase 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322340.1 | 119 | Missense Mutation | AAG,GAG | K,E 33 | NP_001309269.1 | |
NM_001322341.1 | 119 | UTR 5 | NP_001309270.1 | |||
NM_001322342.1 | 119 | Intron | NP_001309271.1 | |||
NM_001322343.1 | 119 | Intron | NP_001309272.1 | |||
NM_001322344.1 | 119 | Missense Mutation | AAG,GAG | K,E 33 | NP_001309273.1 | |
NM_022779.8 | 119 | Missense Mutation | AAG,GAG | K,E 33 | NP_073616.6 | |
NM_138620.1 | 119 | Missense Mutation | AAG,GAG | K,E 33 | NP_619526.1 | |
XM_005272207.4 | 119 | Intron | XP_005272264.1 | |||
XM_006717236.3 | 119 | Missense Mutation | AAG,GAG | K,E 33 | XP_006717299.1 | |
XM_011518921.2 | 119 | Missense Mutation | AAG,GAG | K,E 33 | XP_011517223.1 | |
XM_011518922.2 | 119 | Missense Mutation | AAG,GAG | K,E 33 | XP_011517224.1 |
GTF3C4 - general transcription factor IIIC subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012204.3 | 119 | UTR 5 | NP_036336.2 | |||
XM_017015317.1 | 119 | Intron | XP_016870806.1 | |||
XM_017015318.1 | 119 | Intron | XP_016870807.1 |