Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGCCCCCACCTGGCCACAGCC[A/G]CCTCTCCAGGCTTCCTTGCACACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC5L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARPC5L - actin related protein 2/3 complex subunit 5 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL35 - ribosomal protein L35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR38 - WD repeat domain 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001045476.2 | Intron | NP_001038941.1 | ||||
NM_001276374.1 | Intron | NP_001263303.1 | ||||
NM_001276375.1 | Intron | NP_001263304.1 | ||||
NM_001276376.1 | Intron | NP_001263305.1 | ||||
XM_005251987.3 | Intron | XP_005252044.1 |