Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTAGATGCCACTGTTCTTAGCC[A/T]TGAAACTACATAATATGTTATTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611075 MIM: 611149 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPH5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPH5 - diphthamide biosynthesis 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077394.1 | Intron | NP_001070862.1 | ||||
NM_001077395.1 | Intron | NP_001070863.1 | ||||
NM_015958.2 | Intron | NP_057042.2 | ||||
XM_005270938.1 | Intron | XP_005270995.1 | ||||
XM_005270939.4 | Intron | XP_005270996.1 | ||||
XM_005270941.3 | Intron | XP_005270998.1 | ||||
XM_006710683.3 | Intron | XP_006710746.1 | ||||
XM_011541569.1 | Intron | XP_011539871.1 | ||||
XM_011541570.2 | Intron | XP_011539872.1 | ||||
XM_017001460.1 | Intron | XP_016856949.1 | ||||
XM_017001461.1 | Intron | XP_016856950.1 | ||||
XM_017001462.1 | Intron | XP_016856951.1 |
SLC30A7 - solute carrier family 30 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144884.1 | Intron | NP_001138356.1 | ||||
NM_133496.4 | Intron | NP_598003.2 | ||||
XM_011540779.2 | Intron | XP_011539081.1 | ||||
XM_017000400.1 | Intron | XP_016855889.1 | ||||
XM_017000401.1 | Intron | XP_016855890.1 |