Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGACCCACCATGATCAGTGTG[A/G]TTGTCTCTTGGCTGGTGGCCCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2J1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2J1 - ubiquitin conjugating enzyme E2 J1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016021.2 | 1171 | Silent Mutation | AAC,AAT | N,N 280 | NP_057105.2 | |
XM_011535887.1 | 1171 | Silent Mutation | AAC,AAT | N,N 240 | XP_011534189.1 | |
XM_011535888.2 | 1171 | UTR 3 | XP_011534190.1 |