Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAAAAAGAACAATAAAATGTCA[A/C]ATATTTTTAGAGAAGACATAGTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXTL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXTL2 - exostosin like glycosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033025.2 | Intron | NP_001028197.1 | ||||
NM_001261440.1 | Intron | NP_001248369.1 | ||||
NM_001261441.1 | Intron | NP_001248370.1 | ||||
NM_001261442.1 | Intron | NP_001248371.1 | ||||
NM_001439.3 | Intron | NP_001430.1 | ||||
XM_005270621.1 | Intron | XP_005270678.1 | ||||
XM_011540994.2 | Intron | XP_011539296.1 | ||||
XM_011540995.2 | Intron | XP_011539297.1 | ||||
XM_011540996.2 | Intron | XP_011539298.1 | ||||
XM_017000651.1 | Intron | XP_016856140.1 |