Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGCTGACAACGTCCCCGGGGGC[A/G]CCGCACACACCACACCCACCAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605107 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EDF1 - endothelial differentiation related factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281297.1 | Intron | NP_001268226.1 | ||||
NM_001281298.1 | Intron | NP_001268227.1 | ||||
NM_001281299.1 | Intron | NP_001268228.1 | ||||
NM_003792.3 | Intron | NP_003783.1 | ||||
NM_153200.2 | Intron | NP_694880.1 |
MAMDC4 - MAM domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |