Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCAGGGGCAAAACTGTGAGCT[G/T]GGTAGGAGTTGCAGCAGTAGCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605682 MIM: 602387 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAZ2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAZ2A - bromodomain adjacent to zinc finger domain 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300905.1 | Intron | NP_001287834.1 | ||||
NM_013449.3 | Intron | NP_038477.2 | ||||
XM_011537797.2 | Intron | XP_011536099.1 | ||||
XM_011537798.2 | Intron | XP_011536100.1 | ||||
XM_011537799.2 | Intron | XP_011536101.1 | ||||
XM_011537800.2 | Intron | XP_011536102.1 | ||||
XM_011537801.2 | Intron | XP_011536103.1 | ||||
XM_011537803.2 | Intron | XP_011536105.1 | ||||
XM_011537804.2 | Intron | XP_011536106.1 | ||||
XM_011537805.2 | Intron | XP_011536107.1 | ||||
XM_011537806.2 | Intron | XP_011536108.1 | ||||
XM_011537807.2 | Intron | XP_011536109.1 | ||||
XM_011537809.2 | Intron | XP_011536111.1 | ||||
XM_017018740.1 | Intron | XP_016874229.1 | ||||
XM_017018741.1 | Intron | XP_016874230.1 | ||||
XM_017018742.1 | Intron | XP_016874231.1 |
RBMS2 - RNA binding motif single stranded interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |