Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGAACATCTCACACAGCCTGTT[C/G]ACACAGCTCCTCGAGTTCCTCCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612482 MIM: 603555 | ||||||||||||||||||||
Literature Links: |
RNF43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF43 - ring finger protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305544.1 | 3086 | Missense Mutation | CAA,GAA | Q,E 780 | NP_001292473.1 | |
NM_001305545.1 | 3086 | Missense Mutation | CAA,GAA | Q,E 653 | NP_001292474.1 | |
NM_017763.5 | 3086 | Missense Mutation | CAA,GAA | Q,E 780 | NP_060233.3 | |
XM_011524955.2 | 3086 | Intron | XP_011523257.1 | |||
XM_011524956.2 | 3086 | Intron | XP_011523258.1 | |||
XM_017024800.1 | 3086 | Intron | XP_016880289.1 |
SUPT4H1 - SPT4 homolog, DSIF elongation factor subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPOAP1-AS1 - TSPOAP1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |