Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCTGCCAGGCAGTGGCCCGCCC[A/G]GAGGGAGCTAGGCGGTCCCCACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDX2 - caudal type homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001265.4 | Intron | NP_001256.3 | ||||
XM_011534875.2 | Intron | XP_011533177.1 | ||||
XM_011534876.2 | Intron | XP_011533178.1 | ||||
XM_011534877.2 | Intron | XP_011533179.1 | ||||
XM_011534878.2 | Intron | XP_011533180.1 | ||||
XM_011534879.2 | Intron | XP_011533181.1 |
LINC00543 - long intergenic non-protein coding RNA 543 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |