Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607503 MIM: 614718 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DISP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DISP2 - dispatched RND transporter family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KNSTRN - kinetochore localized astrin/SPAG5 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142761.1 | Intron | NP_001136233.1 | ||||
NM_001142762.1 | Intron | NP_001136234.1 | ||||
NM_033286.3 | Intron | NP_150628.3 |