Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616598 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BORCS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BORCS5 - BLOC-1 related complex subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011520551.2 | 51 | Intron | XP_011518853.1 | |||
XM_011520552.2 | 51 | UTR 5 | XP_011518854.1 | |||
XM_011520553.2 | 51 | UTR 5 | XP_011518855.1 | |||
XM_017018778.1 | 51 | UTR 5 | XP_016874267.1 |
LOH12CR2 - loss of heterozygosity, 12, chromosomal region 2 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MANSC1 - MANSC domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |