Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTAAAGAGCAAAGTAAGAAAAA[G/T]TGAATAAAAAATTCTACGTATATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612500 MIM: 607291 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX52 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX52 - DEAD-box helicase 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291476.1 | Intron | NP_001278405.1 | ||||
NM_007010.4 | Intron | NP_008941.3 | ||||
XM_011524232.2 | Intron | XP_011522534.1 | ||||
XM_011524233.2 | Intron | XP_011522535.1 |
MIR378J - microRNA 378j | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |