Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAGTGCAGAGTGGCTCATGCCCA[C/T]GGTTGACAGCCCATTTACGGCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LNX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LNX2 - ligand of numb-protein X 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153371.3 | 2146 | Missense Mutation | ATG,GTG | M,V 660 | NP_699202.1 | |
XM_011534995.2 | 2146 | Missense Mutation | ATG,GTG | M,V 537 | XP_011533297.1 | |
XM_017020434.1 | 2146 | Missense Mutation | ATG,GTG | M,V 660 | XP_016875923.1 |