Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTGCAGGAGTTGGGGAAGCGGT[C/G]AGCAGTGCCATCCTCTGCTGTCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606610 | ||||||||||||||||||||
Literature Links: |
NSFL1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NSFL1C - NSFL1 cofactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206736.1 | Intron | NP_001193665.1 | ||||
NM_016143.4 | Intron | NP_057227.2 | ||||
NM_018839.4 | Intron | NP_061327.2 |