Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACCATTTCAGGATACACAAGCCC[C/G]CCATTCATTTCCCTCCCTCCCGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611505 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPP - centromere protein P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012267.2 | Intron | NP_001012267.1 | ||||
NM_001286969.1 | Intron | NP_001273898.1 | ||||
NM_001286971.1 | Intron | NP_001273900.1 | ||||
XM_011518685.2 | Intron | XP_011516987.1 | ||||
XM_011518689.1 | Intron | XP_011516991.1 | ||||
XM_017014715.1 | Intron | XP_016870204.1 |
IPPK - inositol-pentakisphosphate 2-kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022755.5 | Intron | NP_073592.1 | ||||
XM_017015041.1 | Intron | XP_016870530.1 |
LOC100128361 - uncharacterized LOC100128361 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |