Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610702 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPA12B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPA12B - heat shock protein family A (Hsp70) member 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197327.1 | Intron | NP_001184256.1 | ||||
NM_001318322.1 | Intron | NP_001305251.1 | ||||
NM_052970.4 | Intron | NP_443202.3 | ||||
XM_011529151.2 | Intron | XP_011527453.1 | ||||
XM_017027632.1 | Intron | XP_016883121.1 | ||||
XM_017027633.1 | Intron | XP_016883122.1 | ||||
XM_017027634.1 | Intron | XP_016883123.1 | ||||
XM_017027635.1 | Intron | XP_016883124.1 |