Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAACATCCCTCATCAGTTCCCCCT[A/G]GATCCCAGCCCGGGGAGGGGGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NECAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NECAB2 - N-terminal EF-hand calcium binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019065.2 | 940 | Intron | NP_061938.2 | |||
XM_011523135.1 | 940 | Intron | XP_011521437.1 | |||
XM_017023317.1 | 940 | Nonsense Mutation | TAG,TGG | *,W 41 | XP_016878806.1 |
OSGIN1 - oxidative stress induced growth inhibitor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |