Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf42 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2orf42 - chromosome 2 open reading frame 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017880.1 | Intron | NP_060350.1 | ||||
XM_005264389.2 | Intron | XP_005264446.1 | ||||
XM_005264390.2 | Intron | XP_005264447.1 | ||||
XM_005264391.2 | Intron | XP_005264448.1 | ||||
XM_011532937.2 | Intron | XP_011531239.1 | ||||
XM_017004398.1 | Intron | XP_016859887.1 | ||||
XM_017004399.1 | Intron | XP_016859888.1 | ||||
XM_017004400.1 | Intron | XP_016859889.1 |