Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603363 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C3orf38 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C3orf38 - chromosome 3 open reading frame 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173824.3 | 436 | Silent Mutation | ACC,ACT | T,T 32 | NP_776185.2 | |
XM_005264745.4 | 436 | Silent Mutation | ACC,ACT | T,T 32 | XP_005264802.1 |
CGGBP1 - CGG triplet repeat binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008390.1 | 436 | Intron | NP_001008391.1 | |||
NM_001195308.1 | 436 | Intron | NP_001182237.1 | |||
NM_003663.3 | 436 | Intron | NP_003654.3 | |||
XM_017007355.1 | 436 | Intron | XP_016862844.1 |
ZNF654 - zinc finger protein 654 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |