Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGGAGTAAATGGGCCTCTGGC[A/G]TGCAGTGGAGAAAGAGCAGGAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601592 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAPSN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAPSN - receptor associated protein of the synapse | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005055.4 | 1510 | UTR 3 | NP_005046.2 | |||
NM_032645.4 | 1510 | UTR 3 | NP_116034.2 | |||
XM_005253042.3 | 1510 | UTR 3 | XP_005253099.1 | |||
XM_005253043.3 | 1510 | UTR 3 | XP_005253100.1 | |||
XM_011520252.1 | 1510 | Missense Mutation | CGC,TGC | R,C 461 | XP_011518554.1 |