Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATGTTATTTGGGATTACGTTG[C/T]AAACTCATTAAACACCAAATTACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609674 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESCO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESCO1 - establishment of sister chromatid cohesion N-acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052911.2 | Intron | NP_443143.2 | ||||
XM_011525798.1 | Intron | XP_011524100.1 | ||||
XM_011525799.2 | Intron | XP_011524101.1 |
GREB1L - growth regulation by estrogen in breast cancer 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |