Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAGTGCCCCAATCCTATTTTCC[A/G]AAGAGGCTTACATACCTGTAAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602345 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPC3 - transient receptor potential cation channel subfamily C member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130698.1 | Intron | NP_001124170.1 | ||||
NM_003305.2 | Intron | NP_003296.1 | ||||
XM_011532217.2 | Intron | XP_011530519.1 | ||||
XM_011532218.2 | Intron | XP_011530520.1 | ||||
XM_017008578.1 | Intron | XP_016864067.1 | ||||
XM_017008579.1 | Intron | XP_016864068.1 |