Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTGCACGCTTCCTTCCACGCTC[C/G]TTGGGACATCGTAGGCATATTCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612303 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRA3 - adhesion G protein-coupled receptor A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145290.3 | 3857 | Missense Mutation | ACG,AGG | T,R 1196 | NP_660333.2 | |
XM_005248137.4 | 3857 | Intron | XP_005248194.1 | |||
XM_011513811.2 | 3857 | Missense Mutation | ACG,AGG | T,R 970 | XP_011512113.1 | |
XM_017007834.1 | 3857 | Missense Mutation | ACG,AGG | T,R 999 | XP_016863323.1 |