Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGGACCCTCTTAGCTCATGGGA[C/T]GACGGCACGACCCCCAGGCCCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613037 MIM: 613036 MIM: 612854 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPP5E - inositol polyphosphate-5-phosphatase E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318502.1 | Intron | NP_001305431.1 | ||||
NM_019892.5 | Intron | NP_063945.2 | ||||
XM_017014926.1 | Intron | XP_016870415.1 |
PMPCA - peptidase, mitochondrial processing alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC16A - SEC16 homolog A, endoplasmic reticulum export factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |