Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602193 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC29A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC29A1 - solute carrier family 29 member 1 (Augustine blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001078175.2 | 380 | Silent Mutation | CCA,CCG | P,P 28 | NP_001071643.1 | |
NM_001078177.1 | 380 | Silent Mutation | CCA,CCG | P,P 28 | NP_001071645.1 | |
NM_001304462.1 | 380 | Silent Mutation | CCA,CCG | P,P 107 | NP_001291391.1 | |
NM_001304463.1 | 380 | Silent Mutation | CCA,CCG | P,P 70 | NP_001291392.1 | |
NM_001304465.1 | 380 | Silent Mutation | CCA,CCG | P,P 54 | NP_001291394.1 | |
NM_001304466.1 | 380 | Silent Mutation | CCA,CCG | P,P 53 | NP_001291395.1 | |
XM_005248876.4 | 380 | Silent Mutation | CCA,CCG | P,P 71 | XP_005248933.1 | |
XM_005248878.3 | 380 | Silent Mutation | CCA,CCG | P,P 28 | XP_005248935.1 | |
XM_005248879.3 | 380 | Silent Mutation | CCA,CCG | P,P 28 | XP_005248936.1 | |
XM_005248880.3 | 380 | Silent Mutation | CCA,CCG | P,P 28 | XP_005248937.1 | |
XM_005248881.3 | 380 | Silent Mutation | CCA,CCG | P,P 28 | XP_005248938.1 | |
XM_005248882.3 | 380 | Silent Mutation | CCA,CCG | P,P 28 | XP_005248939.1 | |
XM_011514341.2 | 380 | Silent Mutation | CCA,CCG | P,P 108 | XP_011512643.1 |
Set Membership: |
DME Validated Inventoried |