Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCTCGAAGCTCTTCTTGCCATG[A/G]TAGGATCCCATGCCGCTGTTCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100660 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ALDH3A1 - aldehyde dehydrogenase 3 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000691.4 | 1183 | Silent Mutation | TAC,TAT | Y,Y 413 | NP_000682.3 | |
NM_001135167.1 | 1183 | Silent Mutation | TAC,TAT | Y,Y 413 | NP_001128639.1 | |
NM_001135168.1 | 1183 | Silent Mutation | TAC,TAT | Y,Y 413 | NP_001128640.1 | |
XM_005256522.1 | 1183 | Silent Mutation | TAC,TAT | Y,Y 530 | XP_005256579.1 | |
XM_005256523.2 | 1183 | Silent Mutation | TAC,TAT | Y,Y 530 | XP_005256580.1 | |
XM_005256524.3 | 1183 | Silent Mutation | TAC,TAT | Y,Y 530 | XP_005256581.1 | |
XM_011523730.1 | 1183 | Silent Mutation | TAC,TAT | Y,Y 340 | XP_011522032.1 | |
XM_011523731.1 | 1183 | Silent Mutation | TAC,TAT | Y,Y 340 | XP_011522033.1 |
Set Membership: |
DME Validated Inventoried |