Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605209 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHFR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHFR - checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161344.1 | 2188 | UTR 3 | NP_001154816.1 | |||
NM_001161345.1 | 2188 | UTR 3 | NP_001154817.1 | |||
NM_001161346.1 | 2188 | UTR 3 | NP_001154818.1 | |||
NM_001161347.1 | 2188 | UTR 3 | NP_001154819.1 | |||
NM_018223.2 | 2188 | UTR 3 | NP_060693.2 |
Set Membership: |
HapMap |