Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179050 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAMD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GRAMD2 - GRAM domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKM - pyruvate kinase, muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206796.2 | Intron | NP_001193725.1 | ||||
NM_001206797.2 | Intron | NP_001193726.1 | ||||
NM_001206798.2 | Intron | NP_001193727.1 | ||||
NM_001206799.1 | Intron | NP_001193728.1 | ||||
NM_001316318.1 | Intron | NP_001303247.1 | ||||
NM_002654.5 | Intron | NP_002645.3 | ||||
NM_182470.3 | Intron | NP_872270.1 | ||||
NM_182471.3 | Intron | NP_872271.1 | ||||
XM_005254443.1 | Intron | XP_005254500.1 | ||||
XM_005254445.4 | Intron | XP_005254502.1 | ||||
XM_006720570.1 | Intron | XP_006720633.1 | ||||
XM_011521670.2 | Intron | XP_011519972.1 | ||||
XM_017022313.1 | Intron | XP_016877802.1 |
Set Membership: |
HapMap |