Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGGAGAAAATGGTAAAACAAC[A/T]CAAGAAATAGGCTCTGCCCTCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605029 | ||||||||||||||||||||
Literature Links: |
KLRF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLRF1 - killer cell lectin like receptor F1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291822.1 | Intron | NP_001278751.1 | ||||
NM_001291823.1 | Intron | NP_001278752.1 | ||||
NM_016523.2 | Intron | NP_057607.1 | ||||
XM_017019414.1 | Intron | XP_016874903.1 | ||||
XM_017019415.1 | Intron | XP_016874904.1 |