Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTCTAGAAGATTTACGGACATC[C/T]CGTCCTTCCCCCTTCTCGCCCCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604935 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMRT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DMRT2 - doublesex and mab-3 related transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130865.2 | Intron | NP_001124337.1 | ||||
NM_006557.6 | Intron | NP_006548.1 | ||||
NM_181872.4 | Intron | NP_870987.2 | ||||
XM_011517687.1 | Intron | XP_011515989.1 | ||||
XM_011517690.2 | Intron | XP_011515992.1 | ||||
XM_011517694.2 | Intron | XP_011515996.1 | ||||
XM_017014213.1 | Intron | XP_016869702.1 | ||||
XM_017014214.1 | Intron | XP_016869703.1 | ||||
XM_017014215.1 | Intron | XP_016869704.1 | ||||
XM_017014216.1 | Intron | XP_016869705.1 |
LINC01230 - long intergenic non-protein coding RNA 1230 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |