Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGAAAAGAGAACAGACAGAAGA[G/T]AAGAGAAGAGAAGGGTATACAGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MORC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MORC4 - MORC family CW-type zinc finger 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085354.2 | 3082 | UTR 3 | NP_001078823.1 | |||
NM_024657.4 | 3082 | UTR 3 | NP_078933.3 | |||
XM_005262190.3 | 3082 | UTR 3 | XP_005262247.1 | |||
XM_006724691.2 | 3082 | UTR 3 | XP_006724754.1 | |||
XM_011531027.2 | 3082 | UTR 3 | XP_011529329.1 | |||
XM_017029844.1 | 3082 | UTR 3 | XP_016885333.1 |