Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTAAAACACCCAAATCATCCGGC[A/G]GGTTACCAGAATGTGGCTGTCTCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606897 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYST PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LYST - lysosomal trafficking regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000081.3 | Intron | NP_000072.2 | ||||
NM_001301365.1 | Intron | NP_001288294.1 | ||||
XM_011544031.1 | Intron | XP_011542333.1 | ||||
XM_011544032.1 | Intron | XP_011542334.1 | ||||
XM_011544033.2 | Intron | XP_011542335.1 | ||||
XM_011544034.1 | Intron | XP_011542336.1 | ||||
XM_011544035.2 | Intron | XP_011542337.1 | ||||
XM_011544036.2 | Intron | XP_011542338.1 | ||||
XM_011544037.2 | Intron | XP_011542339.1 | ||||
XM_011544039.2 | Intron | XP_011542341.1 | ||||
XM_011544040.2 | Intron | XP_011542342.1 | ||||
XM_017000150.1 | Intron | XP_016855639.1 | ||||
XM_017000151.1 | Intron | XP_016855640.1 |