Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTAGTTTTCTATTTTGCAAAC[A/G]GCAAGCCATAGTCACCAAGTTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605269 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CORO1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CORO1C - coronin 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105237.2 | Intron | NP_001098707.1 | ||||
NM_001276471.1 | Intron | NP_001263400.1 | ||||
NM_014325.3 | Intron | NP_055140.1 | ||||
XM_011538125.1 | Intron | XP_011536427.1 | ||||
XM_017019121.1 | Intron | XP_016874610.1 | ||||
XM_017019122.1 | Intron | XP_016874611.1 | ||||
XM_017019123.1 | Intron | XP_016874612.1 |