Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGACCTGGCAGGGCCAGAGAGC[C/T]AGGCCGTCCACCCAGTCCCTACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615104 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCKAP5L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCKAP5L - NCK associated protein 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037806.3 | Intron | NP_001032895.2 | ||||
XM_005269050.3 | Intron | XP_005269107.1 | ||||
XM_006719525.2 | Intron | XP_006719588.1 | ||||
XM_011538610.2 | Intron | XP_011536912.1 | ||||
XM_011538611.2 | Intron | XP_011536913.1 |