Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146691 MIM: 612911 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMPDH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
IMPDH2 - inosine monophosphate dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFAF3 - NADH:ubiquinone oxidoreductase complex assembly factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QRICH1 - glutamine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320580.1 | Intron | NP_001307509.1 | ||||
NM_001320581.1 | Intron | NP_001307510.1 | ||||
NM_001320582.1 | Intron | NP_001307511.1 | ||||
NM_001320583.1 | Intron | NP_001307512.1 | ||||
NM_001320584.1 | Intron | NP_001307513.1 | ||||
NM_001320585.1 | Intron | NP_001307514.1 | ||||
NM_017730.3 | Intron | NP_060200.2 | ||||
NM_198880.2 | Intron | NP_942581.1 | ||||
XM_011533863.1 | Intron | XP_011532165.1 |
Set Membership: |
HapMap Validated |