Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCCGATATCTGTTACAGGATAT[A/G]GAAGCAGCAGAAAAGTCACTACAGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
C3orf14 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
C3orf14 - chromosome 3 open reading frame 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291941.1 | 497 | Missense Mutation | ATA,ATG | I,M 61 | NP_001278870.1 | |
NM_001291942.1 | 497 | Missense Mutation | ATA,ATG | I,M 61 | NP_001278871.1 | |
NM_001291943.1 | 497 | Intron | NP_001278872.1 | |||
NM_020685.4 | 497 | Missense Mutation | ATA,ATG | I,M 61 | NP_065736.1 | |
XM_017006931.1 | 497 | Missense Mutation | ATA,ATG | I,M 61 | XP_016862420.1 |