Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGCCCCACGGTGGGACGCAGGC[A/G]TGACACGGGAAAGTTTCTTGGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605314 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDAC4 - histone deacetylase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006037.3 | 3571 | UTR 3 | NP_006028.2 | |||
XM_006712877.3 | 3571 | UTR 3 | XP_006712940.1 | |||
XM_006712878.3 | 3571 | UTR 3 | XP_006712941.1 | |||
XM_006712879.3 | 3571 | UTR 3 | XP_006712942.1 | |||
XM_006712880.3 | 3571 | UTR 3 | XP_006712943.1 | |||
XM_011512217.2 | 3571 | UTR 3 | XP_011510519.1 | |||
XM_011512218.2 | 3571 | UTR 3 | XP_011510520.1 | |||
XM_011512219.2 | 3571 | UTR 3 | XP_011510521.1 | |||
XM_011512220.2 | 3571 | UTR 3 | XP_011510522.1 | |||
XM_011512221.1 | 3571 | UTR 3 | XP_011510523.1 | |||
XM_011512222.2 | 3571 | UTR 3 | XP_011510524.1 | |||
XM_011512223.2 | 3571 | UTR 3 | XP_011510525.1 | |||
XM_011512224.2 | 3571 | UTR 3 | XP_011510526.1 | |||
XM_011512225.2 | 3571 | UTR 3 | XP_011510527.1 | |||
XM_011512226.2 | 3571 | UTR 3 | XP_011510528.1 | |||
XM_011512227.2 | 3571 | UTR 3 | XP_011510529.1 | |||
XM_011512230.1 | 3571 | UTR 3 | XP_011510532.1 | |||
XM_017005394.1 | 3571 | UTR 3 | XP_016860883.1 | |||
XM_017005395.1 | 3571 | UTR 3 | XP_016860884.1 |