Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCTAATGCCTGTGATTCCTTCA[C/T]TCCCCCACCCCCAGGCCTCTCCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL3 - B-cell CLL/lymphoma 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005178.4 | Intron | NP_005169.2 | ||||
XM_011527197.2 | Intron | XP_011525499.2 | ||||
XM_011527198.2 | Intron | XP_011525500.2 | ||||
XM_011527200.2 | Intron | XP_011525502.1 | ||||
XM_017027109.1 | Intron | XP_016882598.1 | ||||
XM_017027110.1 | Intron | XP_016882599.1 |
MIR8085 - microRNA 8085 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |