Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCCCGTCCCCAGGTGTGCCAC[G/T]GACCACAATGTGGACAACACCACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616626 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CERCAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CERCAM - cerebral endothelial cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286760.1 | 605 | UTR 5 | NP_001273689.1 | |||
NM_016174.4 | 605 | Silent Mutation | ACG,ACT | T,T 69 | NP_057258.3 | |
XM_005252035.1 | 605 | Silent Mutation | ACG,ACT | T,T 69 | XP_005252092.1 | |
XM_011518762.1 | 605 | Silent Mutation | ACG,ACT | T,T 69 | XP_011517064.1 | |
XM_011518763.2 | 605 | UTR 5 | XP_011517065.1 | |||
XM_017014794.1 | 605 | Silent Mutation | ACG,ACT | T,T 69 | XP_016870283.1 |