Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGAATATTCCTGAATGGGCTCA[C/T]GTGGATTTCATCTGGGGTTTGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613923 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD22 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD22 - ankyrin repeat domain 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144590.2 | 3498 | UTR 3 | NP_653191.2 |
LIPM - lipase family member M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128215.1 | 3498 | Silent Mutation | CAC,CAT | H,H 386 | NP_001121687.1 | |
XM_011539748.2 | 3498 | Silent Mutation | CAC,CAT | H,H 393 | XP_011538050.1 | |
XM_011539751.2 | 3498 | Silent Mutation | CAC,CAT | H,H 265 | XP_011538053.1 | |
XM_011539752.2 | 3498 | Silent Mutation | CAC,CAT | H,H 203 | XP_011538054.1 | |
XM_011539753.2 | 3498 | Intron | XP_011538055.1 |
Set Membership: |
HapMap |