Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCGCGTTGATATGATTGGCCGGC[A/G]AATCGTGGTTCTCTTTTCCTCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614258 MIM: 602412 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLR3A - RNA polymerase III subunit A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS24 - ribosomal protein S24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001026.4 | 93 | Intron | NP_001017.1 | |||
NM_001142282.1 | 93 | Intron | NP_001135754.1 | |||
NM_001142283.1 | 93 | Intron | NP_001135755.1 | |||
NM_001142284.1 | 93 | Intron | NP_001135756.1 | |||
NM_001142285.1 | 93 | UTR 5 | NP_001135757.1 | |||
NM_033022.3 | 93 | Intron | NP_148982.1 |