Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGAGGCATACCCACTGGGGCG[C/T]AGGTCTGGGACTCCCCCTGTCCGCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
RAD54L2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RAD54L2 - RAD54-like 2 (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX264 - testis expressed 264 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129884.2 | 56 | Intron | NP_001123356.1 | |||
NM_001243725.1 | 56 | UTR 5 | NP_001230654.1 | |||
NM_001243726.1 | 56 | UTR 5 | NP_001230655.1 | |||
NM_001243727.2 | 56 | Intron | NP_001230656.1 | |||
NM_001278195.1 | 56 | Intron | NP_001265124.1 | |||
NM_015926.5 | 56 | Intron | NP_057010.1 | |||
XM_006713195.3 | 56 | Intron | XP_006713258.1 | |||
XM_006713197.3 | 56 | Intron | XP_006713260.1 | |||
XM_006713198.2 | 56 | Intron | XP_006713261.1 | |||
XM_011533805.2 | 56 | UTR 5 | XP_011532107.1 | |||
XM_017006573.1 | 56 | UTR 5 | XP_016862062.1 | |||
XM_017006574.1 | 56 | Intron | XP_016862063.1 |