Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAACTCCTGTAAGGTACAGAGT[C/T]AGAGATATGACGGTCCCTACAGCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613923 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD22 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD22 - ankyrin repeat domain 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144590.2 | 3630 | UTR 3 | NP_653191.2 |
LIPM - lipase family member M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128215.1 | 3630 | Silent Mutation | GTC,GTT | V,V 342 | NP_001121687.1 | |
XM_011539748.2 | 3630 | Silent Mutation | GTC,GTT | V,V 349 | XP_011538050.1 | |
XM_011539751.2 | 3630 | Silent Mutation | GTC,GTT | V,V 221 | XP_011538053.1 | |
XM_011539752.2 | 3630 | Silent Mutation | GTC,GTT | V,V 159 | XP_011538054.1 | |
XM_011539753.2 | 3630 | Intron | XP_011538055.1 |
Set Membership: |
HapMap |