Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGGGGCGGCAACCGCAGTGACC[A/G]CAGGGCATGGCCGAGTACAGGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600047 MIM: 606533 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABCA2 - ATP binding cassette subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001606.4 | 7560 | UTR 3 | NP_001597.2 | |||
NM_212533.2 | 7560 | UTR 3 | NP_997698.1 | |||
XM_006716996.3 | 7560 | UTR 3 | XP_006717059.1 |
CLIC3 - chloride intracellular channel 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |